missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98778-100ul
This item is not returnable.
View return policy
Description
SGK1 Polyclonal antibody specifically detects SGK1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SGK1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate | |
| EC 2.7.11, EC 2.7.11.1, serine/threonine protein kinase SGK, serine/threonine-protein kinase Sgk1, serum/glucocorticoid regulated kinase, serum/glucocorticoid regulated kinase 1, Serum/glucocorticoid-regulated kinase 1, SGK | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SGK1. | |
| 100 μg | |
| Apoptosis, DNA Repair | |
| 6446 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction