missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SH2D1B Recombinant Protein

Product Code. 18215863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215863 250 μg 250µg
18230653 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18215863 Supplier Novus Biologicals™ Supplier No. NBP260211250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-132 of Human SH2D1B The Recombinant Human SH2D1B Protein is derived from E. coli. The Recombinant Human SH2D1B Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) containing 20% glycerol. containingno Preservative
Gene ID (Entrez) 117157
Name Recombinant SH2D1B Protein
Purification Method >90%, by SDS-PAGE
Quantity 250 μg
Source Baculovirus
Storage Requirements Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1ug of protein (determined by LAL method)
Gene Alias EAT2EAT-2, EWS/FLI1-activated transcript 2, SH2 domain containing 1B, SH2 domain-containing molecule EAT2, SH2 domain-containing protein 1B
Gene Symbol SH2D1B
Product Type Recombinant Protein
Recombinant Recombinant
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.