missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18539-1-AP-20UL
This item is not returnable.
View return policy
Description
Lad (TSAD, TSAd, F2771, SH2D2A, SH2 domain protein 2A) is a 52 kDa cytoplasmic adapter protein that undergoes tyrosine-phosphorylation and influences T cell activation. Lad (SH2D2A) mRNA is present in peripheral blood leukocytes, thymus and spleen, and accumulates in the cytoplasm during T cell activation. The Lad gene maps to chromosome 1q21 in a region where alterations are characteristic to lymphomas.Specifications
| SH2D2A | |
| Polyclonal | |
| Unconjugated | |
| SH2D2A | |
| F2771, SCAP, SH2 domain protein 2A, SH2D2A, TSAd, VRAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9047 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NP31 | |
| SH2D2A | |
| SH2D2A Fusion Protein Ag13355 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction