Learn More
SH3BP1 Monoclonal Antibody (OTI1C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811580
Description
Binds differentially to the SH3 domains of certain proteins of signal transduction pathways. This protein binds preferentially to the ABL1 proto-oncogene, SRC and GRB2. Shows GAP activity for Rac-related proteins but not for Rho- or Ras-related proteins. It inhibits PDGF-induced membrane ruffling mediated by Rac.Specifications
| SH3BP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y3L3 | |
| SH3BP1 | |
| Full length human recombit protein of human SH3BP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C9 | |
| Unconjugated | |
| SH3BP1 | |
| ARHGAP43 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23616 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.