Learn More
Heidolph™ Shaker Vibramax 100
Product Code. 10588751 Shop All Heidolph Instruments Products
Change view
Click to view available options
Orbit:
3 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
10588751 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10588751 Supplier Heidolph™ Supplier No. 5442120000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Compact, space-saving shaker/mixer. The Shaker Vibramax 100 has a load capacity of 2 kg.
  • The 3 mm vibration orbit gives excellent results especially in large vessels – allows for smooth to ridged mixing of viscous media and solids
  • Set and continuously adjust the variable speed on the analog control knob from 150 to 1,350 rpm
  • Optional attachment with tension rollers accepts any glassware size
  • A large selection of attachments is provided for the use of various vessels and sizes
  • An analog process timer allows for unattended operation and can be set from 1 to 120 minutes. When the set time has elapsed an acoustic alarm will sound and the operation stops
TRUSTED_SUSTAINABILITY
Specifications
Orbit 3 mm
Load Bearing Capacity (Metric) 2 kg
Speed Range 150 to 1350 rpm
Electrical Requirements 230 V, 31 W
Product Type Shaker
Load Bearing Capacity (English) 4.4 lb.
Certifications/Compliance DIN EN 60529, IP 30
Dimensions (LxW) Platform 220 x 220 mm
Model Vibramax 100
Includes Timer, overheat protection
Timer Range 1 to 120 min.