Learn More
Memmert™ Shaking Device for Waterbaths
Product Code. 17110415 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
WTB35
WTB50
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17110415 WTB50 Each
17180385 WTB35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17110415 Supplier Memmert™ Supplier No. WTB50SV3550
Currently Unavailable: We have no information when this product will be available.
View alternative products
Shaking device with support frame without gable cover & support basket (mandatory)
  • Requires gable cover
  • Voltage 230 V, 50/60 Hz
  • Order the support basket separately
  • Includes support frame, shaking frequency 35 - 160 strokes per minute (horizontal back/forth movement)
Specifications
For Use With (Equipment) WTB50
Type Shaking Device
Speed 35-160 rpm