Learn More
Epredia™ Shandon™ Eosin-Y Stain
Product Code. 10483750 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1L
4L
Each
Product Type:
Alcoholic Eosin Y Stain
Aqueous Eosin Y Stain
Unit Size:
4 L
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
10483750 4L Alcoholic Eosin Y Stain 4 L
10562614 1L Alcoholic Eosin Y Stain Pack of 2
12532756 1L Aqueous Eosin Y Stain Pack of 2
10188418 Each Aqueous Eosin Y Stain 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10483750 Supplier Epredia™ Supplier No. 6766008
  • ONLINE EXCLUSIVE PRICE
    60.65€
    47.31€ / 4 L
    Save 13.34€
    21% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4L
Product Type Alcoholic Eosin Y Stain
compliance-icons
Product Identifier
  • Shandon Eosin-Y Alcoholic
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.