Learn More
Epredia™ Shandon™ Finesse™ Fine Orienting Head
Product Code. 12658046 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Fine Orienting Head
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12658046 Fine Orienting Head Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12658046 Supplier Epredia™ Supplier No. 77510166
  • ONLINE EXCLUSIVE PRICE
    507.00€
    394.48€ / Pack of 1
    Save 112.52€
    22% Off
Estimated Shipment: 05-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed for the Epredia™ Shandon™ Finesse ME/E Series only, the Epredia™ Finesse Fine Orienting Head is of excellent quality.
  • Allows x-y orientation of the specimen
  • Supplied with the Finesse ME+ and E+ model microtomes only
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fine Orienting Head