Learn More
Epredia™ Glyo-Fixx™
Product Code. 12520067 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
125 x 60mL
5 gal.
125 x 120mL
Product Type:
Fixative
Unit Size:
3.8 L
Each
Pack of 125
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Unit Size
12520067 125 x 120mL Fixative Pack of 125
10249908 5 gal. Fixative Each
10602689 1 gal. Fixative 3.8 L
12500067 125 x 60mL Fixative Pack of 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12520067 Supplier Epredia™ Supplier No. 9990928
  • ONLINE EXCLUSIVE PRICE
    264.00€
    205.65€ / Pack of 125
    Save 58.35€
    22% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixative
Quantity 125 x 120mL