Learn More
Epredia™ Shandon™ Heavy-Duty Scalpel, Belly 2 x 0.5 in. (5.0 x 1.3cm) blade, 7 in. (18.0cm) standard
Product Code. 12637736 Shop All Epredia Products
Change view
Click to view available options
Length (English):
7 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) unitSize
12637736 7 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12637736 Supplier Epredia™ Supplier No. 745
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cut with the standard belly blade of the Epredia™ Shandon™ Heavy-Duty Scalpel.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Product Type Belly Blade
Size 5.0 x 1.3 cm
Length (English) 7 in.