Learn More
Epredia™ Shandon™ Littauer-Liston Bone Cutting Forceps, Box Pivot, 2 Spring, premium, 6 in. (15.2cm)
Product Code. 15380865 Shop All Epredia Products
Change view
Click to view available options
Length (English):
6 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15380865 6 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15380865 Supplier Epredia™ Supplier No. 3120011
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Choose 6 in. (15.2cm) Epredia™ Shandon™ Littauer-Liston Bone Cutting Forceps with box pivot and dual spring for quality and precision.
Features:
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 6 in.