Learn More
Epredia™ Shandon™ Specimen Knife with Ruler, Ratcliffe Pattern, 8 x 1.5 in. (20.3 x 3.8cm) blade, 13 in. (33.0cm) standard
Product Code. 12786747 Shop All Epredia Products
Change view
Click to view available options
Length (English):
13 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
12786747 13 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12786747 Supplier Epredia™ Supplier No. 717
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cut specimens with the Epredia™ Shandon™ Specimen Knife with Ruler, featuring a ruler and Ratcliffe pattern.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Guaranteed to be free from defects in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Length (English) Blade 8 in.
Length (Metric) Blade 20.3 cm
Length (English) 13 in.
Description Ratcliffe Pattern, Blade: 8 x 1.5 in. (20.3 x 3.8 cm) Standard; 13 in. (33.0 cm)