missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ SHARPFLEX Latex Gloves

Product Code. 15856745
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15856745 10 1 pair
15826745 7 1 pair
15836745 8 1 pair
15846745 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15856745 Supplier Honeywell™ Supplier No. 223252510
In Stock
Add to basket
Edge
Add to basket

Honeywell™ SHARPFLEX Latex Gloves for handling of heavy parts with high cut risks in humid and slippery environments.

  • Knitted glove in composite and para-aramid yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The para-aramid yarn assure a resistance to the contact heat and cut risks
  • Plated polyamide inside
  • Offer a greater comfort and limit the contact of the skin with composite fiber
  • Blue crinkled latex coating on palm
  • Latex coating provides better grip and a light thermal protection (up to 250°C) resistance
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN407-04 : Thermal risks
Description Para-aramid and composite, 13 gauge
Color Yellow, Green, Blue
Material Latex
Size 10
Length (Metric) 25 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.