Learn More
Hospidex™ SharpSafe™ Biohazard Carton Bags
Product Code. 11938887
Change view
Click to view available options
Capacity (Metric):
20 L
50 L
Color:
Brown
White
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Unit Size
11938887 50 L Brown Pack of 10
11928887 50 L Brown Pack of 10
11918887 20 L White Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11938887 Supplier Hospidex™ Supplier No. ONDANDAP50
  • ONLINE EXCLUSIVE PRICE
    64.45€
    37.50€ / Pack of 10
    Save 26.95€
    41% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use SharpSafe™ Biohazard Carton Bags for effective waste disposal. These containers facilitate the process that is critical not only to avoid regulatory fines but also to prevent the spread of infectious diseases.
  • Cardboard
  • Various sizes and colors
  • Disposable
Specifications
Color Brown
Disposable Single-use
Material Cardboard
Product Type Biohazard Carton Bag
Capacity (Metric) 50 L
Length (Metric) 260 mm