missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHE Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22902-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes the 54 kDa protein.
SHE, also known as SH2 domain-containing adapter protein E, is a 495 amino acid protein whose molecular weight is 54 kDa. The function of SHE remains unknown.Specifications
| SHE | |
| Polyclonal | |
| Unconjugated | |
| SHE | |
| RP11 350G8.8, SHE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 126669, 214547 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5VZ18, Q8BSD5 | |
| She | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction