missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Human IgG whole molecule, AP, Polyclonal, Secondary Antibody, Abnova™
Specifications
Specifications
| Antigen | Sheep anti-Human IgG whole molecule |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Description | Sheep anti-human IgG recognizes human IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | In 50mM Tris-Cl, 150mM NaCl, 1mM MgCl, 0.1mM ZnCl2, 50% (v/v) glycerol, pH 8.0 (0.1% sodium azide) |
| Host Species | Sheep |
| Immunogen | Human IgG whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?