Learn More
Sheep IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Product Code. 18717828
Change view
Click to view available options
Quantity:
125 mL
25 mL
250 mL (2 x 125 mL)
50 mL (2 x 25 mL)
Unit Size:
125mL
250mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18717828 250 mL (2 x 125 mL) 250mL
18785808 25 mL 25mL
18723579 50 mL (2 x 25 mL) 50mL
18784098 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18717828 Supplier R&D Systems Supplier No. VC006250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Donkey Polyclonal Antibody
IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
Dilution Immunohistochemistry
Formulation Supplied as a solution in PBS with preservative.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Donkey
Immunogen Sheep IgG
Purification Method Affinity Purified
Quantity 250 mL (2 x 125 mL)
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects heavy ( gamma ) chains on sheep IgG and light chains on all sheep immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin sheep serum proteins or IgG from human, mouse or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
Target Species Sheep
Content And Storage 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG