missing translation for 'onlineSavingsMsg'
Learn More

Justrite Shelves for EN Cabinet 226-EN 2903

Additional shelf accessory for EN Cabinets 226-EN. Choice of installed or pull out shelves in stainless steel or epoxy coated finishing.

Brand:  Justrite JCB60MY000

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 17722378

  • 125.00€ / Each
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Choice of stainless steel or epoxy coated shelves
  • Choice of installed or pull out shelves
Specifications

Specifications

Stainless Steel
Shelf
Steel
42 cm
Installed
49 cm
EN Cabinet 226-EN 2903
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions