Learn More
Witeg™ Short Adapter
Join objects with different diameter connectors with this adapter. Witeg™ Short Adapter is designed with ST male and female connection joints.
Brand: Witeg™ 0 540 005
Additional Details : Weight : 0.80000kg
Description
- Short length.
- Socket and cone of varying ST.
Specifications
DIN 12257 | |
Glass | |
Socket ST: 29/32 Cone ST: 19/26. |