missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHP-2/PTPN11 Antibody (7A1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26358-50ul
This item is not returnable.
View return policy
Description
SHP-2/PTPN11 Monoclonal antibody specifically detects SHP-2/PTPN11 in Human samples. It is validated for Western Blot, ELISA
Specifications
| SHP-2/PTPN11 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 7A1 | |
| Western Blot 1:500-1:5000, ELISA | |
| BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP-1D, PTP-2C, PTP2CNS1, Shp2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
| A synthesized peptide derived from Human SHP-2/PTPN11 [UniProt Q06124] | |
| 50 μL | |
| Angiogenesis, Cancer, Cell Biology, Lipid and Metabolism, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| 5781 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction