Learn More
SHPRH Monoclonal Antibody (OTI6D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501465S
Encompass_Preferred
 View more versions of this product
Product Code. 17544321
  • 242.00€ / 30µL
Description
SHPRH Monoclonal antibody specifically detects SHPRH in Human samples. It is validated for Western Blot
Specifications
Specifications
SHPRH
Monoclonal
0.97 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P32004
Mouse
Affinity chromatography
RUO
3897
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI6D9
Unconjugated
L1CAM
2610103K11Rik; AA450458; AU024614; bA545I5.2; BC006883; D230017O13Rik; E130018M05; E3 ubiquitin-protein ligase SHPRH; KIAA2023; RING-type E3 ubiquitin transferase SHPRH; Shprh; SNF2 histone linker PHD RING helicase; SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase; SNF2, histone-linker, PHD and RING finger domain-containing helicase
Full length human recombinant protein of human L1CAM produced in HEK293T cell.