Learn More
Berthold Technologies Sideview Kit (IK Version)
Product Code. 31777399 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31777399 Imaging System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31777399 Supplier Berthold Technologies Supplier No. 74647
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
To mount the camera on the side of the imaging chamber in NightSHADE imaging systems.
  • The default camera position in the NightSHADE in vivo imaging system (plant configuration) is on top of the imaging chamber
  • Lateral camera mounting enables side-view imaging
  • Allows seedlings to remain upright in a natural vertical orientation
  • Enables imaging of the entire plant, from leaves to roots
Specifications
Type Connection Kit
For Use With (Application) Plant Imaging
For Use With (Equipment) Imaging System