missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-2/CD22 Antibody (308501), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2296D
This item is not returnable.
View return policy
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Siglec-2/CD22 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 308501 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Siglec-2/CD22 | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology | |
| 933 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction