Learn More
SIGN-R1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547210
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) SIGNR3 and rmSIGNR4 is observed, and approximately 10% cross-reactivity with recombinant human DC-SIGNR and rmDC-SIGN is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
SIGN-R1 Polyclonal antibody specifically detects SIGN-R1 in Mouse samples. It is validated for Neutralization, Western Blot.Specifications
SIGN-R1 | |
Polyclonal | |
Unconjugated | |
Cd209a | |
1810030I22Rik; CD209; CD209 antigen-like protein A; CD209 antigen-like protein B; Cd209a; CD209a antigen; CD209a molecule; Cd209b; CD209b antigen; CDSIGN; CIRE; DC SIGNR1; Dcsign; DC-SIGN; DC-SIGN1; DC-SIGNR1; DC-SIGN-related protein 1; Dendritic cell-specific ICAM-3-grabbing non-integrin; MGC130443; mSIGNR1; OtB7; SIGNR1; SIGN-R1; SIGNR5 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
170786, 69165 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8CJ91, Q91ZX1 | |
Cd209a, Cd209b | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR1 Ser76-Gly325 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |