missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGN-R4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547153
Additional Details : Weight : 0.00010kg
Description
In Western blots, approximately 20% cross-reactivity with recombinant mouse SIGNR1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
SIGN-R4 Polyclonal antibody specifically detects SIGN-R4 in Mouse samples. It is validated for Western Blot.Specifications
SIGN-R4 | |
Polyclonal | |
Unconjugated | |
Cd209e | |
CD209; CD209 antigen-like protein E; Cd209e; CD209e antigen; DC-SIGNR4; DC-SIGN-related protein 4; mSIGNR4; SIGNR4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
170780 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q91ZW7 | |
Cd209e | |
Mouse myeloma cell line NS0-derived recombinant mouse SIGNR4/CD209e Ser41-Trp208 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |