Description
- Clever way to change disposal container without interrupting the inflow
- LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
- When container is changed, alert can be muted via new touchpad
Specifications
Specifications
| For Use With (Equipment) | SCAT™ Sensors and Safety Waste Caps |
| Length (English) | 7.1 in. |
| Length (Metric) | 55 mm |
| Plug Type | UK-style |
| Size | 180 x 105 x 55 mm |
| Voltage | 24 V |
| Amperage | 2 A |
| For Use With (Application) | Monitoring Filling Levels |
| Height (Metric) | 105 mm |
| Includes | 5 Sensor Inputs and 6 Potential-free Outputs |
| Show More |