Learn More
Silencer™ siRNA Library, In Plate
Product Code. 10288794
Change view
Click to view available options
Quantity:
1 μmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10288794 1 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10288794 Supplier Invitrogen Supplier No. AM81810
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Silencer™ siRNA Library, In Plate
Silencer™ siRNA Libraries feature combinations of Silencer™ Pre-designed siRNAs, and when available, Silencer™ Validated siRNAs, in 96-well or 384-well plates.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Library SiRNA, 1μmol, Powder, Room Temperature Storage, In Plate, Custom, feature Combinations of Silencer Pre-designed siRNAs and When Available, Silencer Validated siRNAs, in 96-well or 384-well Plates
Product Type Library SiRNA
Quantity 1 μmol
Product Title
Select an issue
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.