Learn More
Macherey-Nagel™ Standard SIL G Silica Layers on Glass Plates
Product Code. 10308221 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
50 x 100 mm
50 x 200 mm
50x200mm
100 x 100 mm
100 x 200 mm
200 x 200 mm
200 x 200 cm
Quantity:
100 Pk.
12 Pk.
15 Pk.
20 Pk.
25 Pk.
25/Pk.
50 Pk.
Thickness (Metric):
0.25 mm
0.5 mm
1 mm
2 mm
Fluorescent Indicator:
UV254
UV254 + UV366
Unit Size:
Pack of 100
Pack of 12
Pack of 15
Pack of 20
Pack of 25
Pack of 50
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Thickness (Metric) Fluorescent Indicator unitSize
10308221 100 x 100 mm 25 Pk. 0.25 mm UV254 Pack of 25
10469941 100 x 200 mm 50 Pk. 0.25 mm - Pack of 50
10649391 100 x 200 mm 50 Pk. 0.25 mm UV254 + UV366 Pack of 50
11857043 100 x 200 mm 50 Pk. 0.25 mm UV254 Pack of 50
10371102 200 x 200 mm 25 Pk. 0.25 mm UV254 Pack of 25
10015681 200 x 200 mm 25/Pk. 0.25 mm - Pack of 25
10023021 200 x 200 mm 12 Pk. 2 mm UV254 Pack of 12
10055711 200 x 200 cm 25 Pk. 0.25 mm UV254 + UV366 Pack of 25
10698811 200 x 200 mm 20 Pk. 0.5 mm - Pack of 20
10609011 200 x 200 mm 15 Pk. 1 mm - Pack of 15
10184851 200 x 200 mm 12 Pk. 2 mm - Pack of 12
10576701 50 x 100 mm 50 Pk. 0.25 mm - Pack of 50
10575571 50 x 100 mm 50 Pk. 0.25 mm UV254 Pack of 50
10388071 50x200mm 100 Pk. 0.25 mm - Pack of 100
10716152 50 x 200 mm 100 Pk. 0.25 mm UV254 Pack of 100
12770154 50 x 200 mm 100 Pk. 0.25 mm UV254 + UV366 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 10308221 Supplier Macherey-Nagel™ Supplier No. 809020
Add to basket
Add to basket
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.
Glass TLC plates, SIL G/UV 254.
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications
Material Silica 60
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 25 Pk.
Type Glass Plate
Fluorescent Indicator UV254
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.25 mm
Dimensions (L x W) 100 x 100 mm