Learn More
Hirschmann™ Silicone Knobs Lid
Product Code. 10564252
Change view
Click to view available options
Material:
Silicone
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10564252 Silicone Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10564252 Supplier Hirschmann™ Supplier No. 9240796
  • ONLINE EXCLUSIVE PRICE
    94.00€
    58.41€ / Pack of 1
    Save 35.59€
    37% Off
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Silicone lid for Hirschmann™ 96-well plates, pierceable.
  • Made of silicone
Specifications
Material Silicone
Product Type Knobs Lid
Plate Compatibility 96-well Plate