Learn More
De Buyer™ Silicone Baking Mats
Product Code. 11910495
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11910495 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11910495 Supplier De Buyer™ Supplier No. 4931.40N
  • ONLINE EXCLUSIVE PRICE
    60.90€
    22.30€ / Pack of 1
    Save 38.60€
    63% Off
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Order Info
LengthMetric
Specifications
Product Type Baking Mat
Material Silicone
For Use With (Application) Used for hot or cold work to replace disposable baking paper
For Use With (Equipment) Used as a support for baking, refrigerating, deep-freezing and blast-freezing
Height (Metric) 2 mm
Length (Metric) 400 mm
Quantity Each
Temperature Range (Metric) −40 to 250°C
Width (Metric) 300 mm