Learn More
Thermo Scientific™ Silicone-free Viton™ Door Sealing for Thermo Scientific™ Heratherm™ Incubators and Ovens
Product Code. 12625326 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Heratherm 400l Models
Heratherm 750l Models
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12625326 Heratherm 750l Models Each
12615326 Heratherm 400l Models Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12625326 Supplier Thermo Scientific™ Supplier No. 50135870
  • ONLINE EXCLUSIVE PRICE
    1735.00€
    1640.00€ / Pack of 1
    Save 95.00€
    5% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Thermo Scientific™ Heratherm™ Incubators and Ovens
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Heratherm 750l Models
Type Door Seal