missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HotGrip™ Hand Protectors

Product Code. 10700034 Shop All Bürkle Products
Change view
Click to view available options
Color:
Red
White
For Use With (Application):
Protects The Whole Hand From Burns and Freezing
Protects Thumb and 1 Finger From Burns and Freezing
Protects Thumb and 2 to 3 Fingers From Burns and Freezing
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Application) unitSize
10700034 White Protects Thumb and 2 to 3 Fingers From Burns and Freezing Each
10074112 Red Protects The Whole Hand From Burns and Freezing Each
10185322 White Protects Thumb and 1 Finger From Burns and Freezing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10700034 Supplier Buerkle™ Supplier No. 97680003
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from silicone rubber for extreme thermal resistance. Buerkle™ HotGrip™ Hand Protectors protect fingers and hands from burns and freezing.

  • Resistant to temperatures from –50 to +200°C (briefly up to +250°C)
  • Deep frozen, hot or boiling substances and apparatus can be safely handled and positioned
  • Sililcone rubber is extremely elastic and slip-proof; ideal for smooth surfaces
  • Secure grip 
TRUSTED_SUSTAINABILITY

Specifications

Material Silicone Rubber
Color White
For Use With (Application) Protects Thumb and 2 to 3 Fingers From Burns and Freezing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.