Learn More
Uvex Silv-Air Classic 2310 Planet Mask
Product Code. 17935456 Shop All Uvex Products
Change view
Click to view available options
Filter Type:
FFP3
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type Unit Size
17935456 FFP3 Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17935456 Supplier Uvex Supplier No. 8762313
  • ONLINE EXCLUSIVE PRICE
    76.00€
    66.80€ / Pack of 15
    Save 9.20€
    12% Off
View all promotions
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The uvex silv-Air c product range consists of both classic folding and cup-style respirators that fit a wide variety of facial shapes.
The headband strap and integrated sealing lips in all protection classes make it easy to fit these masks correctly – and to wear them comfortably for long periods.
  • Particle-filtering preformed mask with exhalation valve for larger facial shapes
  • Valve and headband holder made of partly recycled material
  • Comfort features
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Very soft, all-round sealing lip for optimum wearer comfort and reliably tight fit
  • Exhalation valve to reduce exhalation resistance as well as heat and moisture under the mask
  • Ergonomically adapted specifically to larger facial shapes
  • FFP3 respirator compliant with EN 149:2001+A1:2009
  • D certification (mask has passed the dolomite dust test): excellent breathability even with high levels of dust
  • Not reusable
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type P - Oil-Proof
Color White
Design Type Cup/Molded
Disposable Reusable
Filter Type FFP3
Material Polypropylene, Polyester
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Sterility Sterile