missing translation for 'onlineSavingsMsg'
Learn More

Silver Bromide, TRC

Product Code. 30419953
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419953 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419953 Supplier Toronto Research Chemicals Supplier No. TRCS465055100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver Bromide
CAS 7785-23-1
Quantity 100 mg
Molecular Formula AgBr
Synonym Argentous Bromide, Silver Monobromide, Silver(I) Bromide
InChI Formula InChI=1S/Ag.BrH/h;1H/q+1;/p-1
SMILES [Br-].[Ag+]
IUPAC Name silver;bromide
Molecular Weight (g/mol) 187.77
Formula Weight 185.8234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.