missing translation for 'onlineSavingsMsg'
Learn More

Silver Carbonate, TRC

Product Code. 30427989
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30427989

Brand: Toronto Research Chemicals TRCS46503525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver Carbonate
CAS 534-16-7
Molecular Formula Ag2CO3
Quantity 25 g
Synonym Disilver carbonate, Silver(I) carbonate, Carbonic Acid Disilver(1+) Salt
InChI Formula InChI=1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
SMILES [Ag]OC(=O)O[Ag]
IUPAC Name argentiooxycarbonyloxysilver
Molecular Weight (g/mol) 275.75
Formula Weight 273.7949
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.