missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Carbonate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Silver Carbonate |
| CAS | 534-16-7 |
| Molecular Formula | Ag2CO3 |
| Quantity | 5 g |
| Synonym | Disilver carbonate, Silver(I) carbonate, Carbonic Acid Disilver(1+) Salt |
| InChI Formula | InChI=1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2 |
| SMILES | [Ag]OC(=O)O[Ag] |
| IUPAC Name | argentiooxycarbonyloxysilver |
| Molecular Weight (g/mol) | 275.75 |
| Formula Weight | 273.7949 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction