Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >100°C |
| Color | Black to Gray |
| Physical Form | Powder |
| Quantity | 10 g |
| Linear Formula | AgO |
| Fieser | 02,369; 04,431; 06,518; 07,322; 09,412; 10,352; 11,469 |
| Merck Index | 15, 8661 |
| Solubility Information | Solubility in water: insoluble in water |
| Formula Weight | 123.87 |
| Percent Purity | 85.5 to 88% Ag |
| Show More |
Safety and Handling
- Silver(II) oxide
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.