missing translation for 'onlineSavingsMsg'
Learn More

Silver Iodide, 99%, Honeywell™ Fluka™

Product Code. 15652250 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15652250

Brand: Honeywell-Fluka 22682325G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

0.99

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-96-2
Molecular Formula AgI
Molecular Weight (g/mol) 234.773
MDL Number MFCD00003412
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
PubChem CID 24563
IUPAC Name iodosilver
SMILES [Ag]I

Specifications

Color Yellow
Physical Form Solid
Quantity 25 g
UN Number UN3077
Formula Weight 234.77g/mol
Percent Purity 99%
Packaging Glass Bottle
Density 5.68g/mL at 25°C
Chemical Name or Material Silver Iodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.