missing translation for 'onlineSavingsMsg'
Learn More

Silver, solder alloy, 3.0mm (0.13in) dia

Product Code. 11399437
Change view
Click to view available options
Quantity:
50 g
250 g
454 g
Unit Size:
250g
454g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399437 250 g 250g
11319447 50 g 50g
15411697 454 g 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399437 Supplier Thermo Scientific Alfa Aesar Supplier No. 041030.30
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver, solder Alloy
Name Note 3.0mm (0.13 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Ag:Sn; 3.5:96.5 wt%
Quantity 250 g
MDL Number MFCD00801108

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.