missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate, TRC

Product Code. 30240652
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240652 5 g 5g
30240550 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240652 Supplier Toronto Research Chemicals Supplier No. TRCS4650605G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2O4S
Molecular Weight (g/mol) 311.8
Synonym Disilver Monosulfate, Disilver Sulfate, Disilver(1+) Sulfate, Silver SulfateSilver Sulfate (Ag2SO4), Sulfuric Acid Disilver Salt, Sulfuric Acid Silver Salt (1:2);
IUPAC Name disilver;sulfate
SMILES [Ag+].[Ag+].[O-]S(=O)(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
Formula Weight 309.7619
Chemical Name or Material Silver Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.