Learn More
Simethicone, TRC
Product Code. 30009759
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009759 50 g 50g
17978304 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009759 Supplier Toronto Research Chemicals Supplier No. TRCS47560050G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8050-81-5
Molecular Formula (C2H6OSi)n•SiO2
Molecular Weight (g/mol) 296.62
Synonym Aligest Plus, Antifoam A, DC Antifoam A, Dow Antifoam A, Dow Corning Antifoam A, Espumisan, KS 66, Mylicon, Q 7-2587, Sentry Simethicone GS, Silfoam SE 2, Simiticone, SonoRx;
SMILES CO[Si](C)(C)O[Si](C)(C)C.O=[Si]=O
Specifications
Quantity 50 g
InChI Formula InChI=1S/C6H18O2Si2.O2Si/c1-7-10(5,6)8-9(2,3)4;1-3-2/h1-6H3;
Formula Weight 296.0752
Chemical Name or Material Simethicone