missing translation for 'onlineSavingsMsg'
Learn More

Sin Nombre Virus Glycoprotein 2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41256-0.025mg

 View more versions of this product

Product Code. 18659997

  • 168.00€ / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sin Nombre Virus Glycoprotein 2 Polyclonal specifically detects Sin Nombre Virus Glycoprotein 2 in Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sin Nombre Virus Glycoprotein 2
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
GPC
Antibody was raised against a 16 amino acid peptide near the center of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 670 - 720 of Sin Nombre Virus Glycoprotein 2. Amino Acid Squence: YSYRRKLVNPANQEET
Affinity Purified
RUO
Virus
IgG
ELISA
1 mg/mL
ELISA 1 μg/mL
Envelope polyprotein, GPC, M polyprotein, SNVsMgp1
Rabbit
126 kDa
0.025 mg
Primary
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.