Learn More
- Home
- Products
- Furniture
- Desks and Tables
- Laboratory Benches and Tables
- Köttermann Single Bench
Köttermann Single Bench
Product Code. 17372189
Change view
Click to view available options
Dimensions (D x W x H):
600 x 900 x 750 mm
600 x 900 x 900 mm
750 x 900 x 750 mm
750 x 900 x 900 mm
600 x 1200 x 750 mm
600 x 1200 x 900 mm
600 x 1500 x 750 mm
600 x 1500 x 900 mm
750 x 1200 x 750 mm
750 x 1200 x 900 mm
750 x 1500 x 750 mm
750 x 1500 x 900 mm
Top Style:
Melamine TopResist Worktop
Trespa TopLab PLUS Worktop
Unit Size:
Each
24 product options available for selection
| Product Code. | Dimensions (D x W x H) | Top Style | unitSize |
|---|---|---|---|
| 17372189 | 600 x 900 x 750 mm | Melamine TopResist Worktop | Each |
| 17302219 | 600 x 900 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17302199 | 600 x 900 x 900 mm | Melamine TopResist Worktop | Each |
| 17332219 | 600 x 900 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17382189 | 600 x 1200 x 750 mm | Melamine TopResist Worktop | Each |
| 17312219 | 600 x 1200 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17312199 | 600 x 1200 x 900 mm | Melamine TopResist Worktop | Each |
| 17342219 | 600 x 1200 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17392189 | 600 x 1500 x 750 mm | Melamine TopResist Worktop | Each |
| 17322219 | 600 x 1500 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17322199 | 600 x 1500 x 900 mm | Melamine TopResist Worktop | Each |
| 17352219 | 600 x 1500 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17332199 | 750 x 900 x 750 mm | Melamine TopResist Worktop | Each |
| 17362219 | 750 x 900 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17362199 | 750 x 900 x 900 mm | Melamine TopResist Worktop | Each |
| 17392219 | 750 x 900 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17342199 | 750 x 1200 x 750 mm | Melamine TopResist Worktop | Each |
| 17372219 | 750 x 1200 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17372199 | 750 x 1200 x 900 mm | Melamine TopResist Worktop | Each |
| 17302229 | 750 x 1200 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17352199 | 750 x 1500 x 750 mm | Melamine TopResist Worktop | Each |
| 17382219 | 750 x 1500 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17382199 | 750 x 1500 x 900 mm | Melamine TopResist Worktop | Each |
| 17312229 | 750 x 1500 x 900 mm | Trespa TopLab PLUS Worktop | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
This item is not returnable.
View return policy
Product Code.
17372189
Supplier
Köttermann
Supplier No.
50500129
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
This item is not returnable.
View return policy
Description
- Available for sitting and standing work height
- Stationary
Specifications
Specifications
| Dimensions (D x W x H) | 600 x 900 x 750 mm |
| No. of Shelves | 1 |
| Top Style | Melamine TopResist Worktop |