missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SIRP alpha/CD172a Antibody (Hospital for Sick Children patent anti-SIRPA), Novus Biologicals™

Human Monoclonal Antibody

213.00€ - 920.00€

Specifications

Antigen SIRP alpha/CD172a
Clone Hospital for Sick Children patent anti-SIRPA
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30045708
View Documents
Novus Biologicals™
NBP3-28670-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044579
View Documents
Novus Biologicals™
NBP3-28670-1mg
1 mg
920.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045533
View Documents
Novus Biologicals™
NBP3-28670-100ug
100 μg
338.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SIRP alpha/CD172a Monoclonal antibody specifically detects SIRP alpha/CD172a in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

SIRP alpha/CD172a
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
GPCR, Protein Phosphatase, Signal Transduction
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1
SIRPa/ CD172a
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Hospital for Sick Children patent anti-SIRPA
Monoclonal
Purified
RUO
Human
P78324
140885
IgG4
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.