missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP BETA 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11811-1-AP-20UL
This item is not returnable.
View return policy
Description
SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.Specifications
| SIRP BETA 1 | |
| Polyclonal | |
| Unconjugated | |
| SIRPB1 | |
| CD172b, SIRP BETA 1, SIRPB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10326 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00241 | |
| SIRPB1 | |
| SIRP BETA 1 Fusion Protein Ag2478 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction