missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIRPB2 Monoclonal Antibody (1008709)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA562063

Product Code. 30534472

  • 399.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIRPB2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q5JXA9
SIRPB2
HEK293 human embryonic kidney cell line, HEK293 derived human SIRP beta 2, Gln33-Gly287.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry
1008709
Unconjugated
SIRPB2
dJ776F14.2; Protein tyrosine phosphatase non-receptor type substrate 1-like 3; protein tyrosine phosphatase non-receptor type substrate protein; protein tyrosine phosphatase, non-receptor type substrate 1-like 3; PTPN1L; PTPNS1L3; signal regulatory protein beta 2; signal-regulatory protein beta 2; signal-regulatory protein beta-2; SIRPB2; SIRP-beta-2
Mouse
Protein A/G
RUO
284759
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.