missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SJ 3149
Click to view available options
:
5 mg
Description
SJ 3149 is a potent and selective CK1α molecular glue Degrader (DC50 = 3.7 nM; Dmax 95% in MOLM-13 cells). SJ 3149 induced near-complete degradation of the CK1α protein (DC50 = 6 nM at 4 hours). Potently inhibits the viability of MOLM-13 cells (IC50 = 13 nM). SJ 3149 displays a broad antiproliferative activity profile on a panel of acute leukemia and solid tumor cell lines. In vivo, SJ 3149 degrades CK1α protein levels in NSG mice transplanted with MOLM-13 cells. SJ 3149 shows 76% orally bioavailability.
Specifications
Specifications
| Chemical Name or Material | 3-(5-(Benzo[d]isoxazol-3-ylamino)-1-oxoisoindolin-2-yl)piperidine-2,6-dione |
| CAS | 3026986-17-1 |
| Quantity | 5 mg |
| Target | Molecular Glues |
| Molecular Formula | C20H16N4O4 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction