Learn More
Lonza™ Skeletal Muscle Cell Growth Media-2 Bulletkit
Description
Order Info
dimProductType
Specifications
Specifications
| Product Type | Growth Medium |
| Quantity | 1 Kit |
| Content And Storage | 500mL SkBM-2 basal medium plus CC-3244 singlequots kit to formulate SkGM-2 medium (growth medium) |