missing translation for 'onlineSavingsMsg'
Learn More

SKF-525A Hydrochloride, TRC

Product Code. 30012850
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012850 100 mg 100mg
30011415 250 mg 250mg
30003926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012850 Supplier Toronto Research Chemicals Supplier No. TRCS550000100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-68-0
Molecular Formula C23 H31 N O2 . Cl H
Molecular Weight (g/mol) 389.96
Synonym Benzeneacetic acid, α-phenyl-α-propyl-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzeneacetic acid, α-phenyl-α-propyl-, 2-(diethylamino)ethyl ester, hydrochloride (9CI), Valeric acid, 2,2-diphenyl-, 2-(diethylamino)ethyl ester hydrochloride (6CI,8CI), 2-(Diethylamino)ethyl 2,2-diphenylvalerate hydrochloride, 2'-Diethylaminoethyl 2,2-diphenylpentanoate hydrochloride, Proadifen hydrochloride, RP 5171, SKF 525, SKF 525A, U 5446, β-Diethylaminoethyl diphenylpropylacetate hydrochloride
IUPAC Name 2-(diethylamino)ethyl 2,2-diphenylpentanoate;hydrochloride
SMILES Cl.CCCC(C(=O)OCCN(CC)CC)(c1ccccc1)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H31NO2.ClH/c1-4-17-23(20-13-9-7-10-14-20,21-15-11-8-12-16-21)22(25)26-19-18-24(5-2)6-3;/h7-16H,4-6,17-19H2,1-3H3;1H
Formula Weight 389.2122
Percent Purity >95%
Chemical Name or Material Proadifen Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.