Learn More
Novus Biologicals™ Slap Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30839-25ug
This item is not returnable.
View return policy
Description
Slap Polyclonal antibody specifically detects Slap in Human samples. It is validated for Western Blot
Specifications
| Slap | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| hSLAP, SLA1, SLAP, SLAP1, SLAP-1, src-like-adapter, Src-like-adapter protein 1, Src-like-adaptor | |
| Synthetic peptide derived from the human Slap protein. | |
| 25 μg | |
| Immunology | |
| 6503 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |