Learn More
Novus Biologicals™ Slap Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30839-25ug
 View more versions of this product
Product Code. 30067157
  • 213.00€ / 25µg
Description
Slap Polyclonal antibody specifically detects Slap in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Slap
Polyclonal
Western Blot : 5-10 μg/mL
hSLAP, SLA1, SLAP, SLAP1, SLAP-1, src-like-adapter, Src-like-adapter protein 1, Src-like-adaptor
Synthetic peptide derived from the human Slap protein.
25 μg
Immunology
6503
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation